Waring BHL240 Blender motor base for one-liter container; explosion proof; 240V

Representative image only.
Waring Mfr # BHL240 – Item # EW-04257-55
Explosion-proof motors allow safe mixing of volatile substances in hazardous locations

$11,900.00USD / Each

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (L)1
  • Max RPM18500
  • Motor TypeExplosion Proof
  • Power (VAC)240
  • Power (amps)4
  • Power (Hz)50/60
  • ColorWhite
  • DescriptionBlender motor base for one-liter container; explosion proof; 240V

More About this Item

Note: blenders must be installed by a qualified electrician to preserve their explosion-proof capability.

One-liter blender base is UL-listed for use in Class 1, Group D hazardous locations; requires explosion-proof switch 04257-60 and 1-liter stainless steel container 04257-70.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-847-549-7600
Live chat