This item is now discontinued. Please browse similar items
Wave Biotech HLTS-II Tube Sealer, Hot Lips, 0.25 To 1.25"od,115/230 VAC
Representative image only.
Wave Biotech Mfr # HLTS-II – Item # EW-06480-30
Securely seal any thermoplastic tubing from 1/4” to 1-1/4” OD
  • Works with liquid-filled tubing — no need to drain before use
  • Sealer only touches outside of tubing, while inside remains contamination-free
  • One-button control provides simple, error-free operation
SPECS & DESCRIPTION
Specifications & Description
  • Max Tubing OD (in)1 1/4
  • DescriptionTube Sealer, Hot Lips, 0.25 To 1.25"od,115/230 VAC
Key Features
  • Works with liquid-filled tubing — no need to drain before use
  • Sealer only touches outside of tubing, while inside remains contamination-free
  • One-button control provides simple, error-free operation
More About this Item
Securely seal a broad range of tubing sizes and materials with this flexible tube sealer. Compatible with all thermoplastics, including Tygon®, C-Flex®, PharMed® BPT and vinyl, it forms a half-inch wide seal that can only be opened when cut, preventing accidental leakage or contact with foreign materials. Compact, lightweight, and versatile, the flexible tube sealer is suitable for a broad range of applications, from sealing media bag inlet and outlet tubing to closing off fermentor or bioprocess vessel sampling ports. Works with Masterflex pump tubing as well as general purpose transfer tubing.
To seal tubing, simply select tubing material, ID, and OD from the pre-programmed LCD menu, insert tubing in the holder, and push start button. The sealer automatically applies the optimum temperature, pressure, and cooling cycle, then opens when seal is set. The display provides continuous feedback during operation, while a PC port allows you to update programs and download new tubing options. Programming lockout prevents unauthorized access.
Flexible tubing sealer can be used in conjunction with sterile tubing fuser 06480-50 to join two liquid-filled sections of tubing. After sealing tubing, transfer to the sterile tubing fuser to connect aseptically without releasing fluid into the fuser or surrounding environment. Please specify voltage setting when ordering.