Weiss DUGY2-0400-2L Pressure Gauge 0-400 PSI, 1/2" NPT(M); 316 SS Sensor/ 316 SS Socket
Representative image only.
Weiss Mfr # DUGY3-400-2L – Item # EW-68900-57
The first-ever "solar only" digital pressure gauge
  • Large display
  • <10 Lux rating- only one foot-candle
  • User selectable functions
  • IP65 weatherproof housing
  • ASME B40.7 conformance
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Accuracy±0.5% full-scale
  • Min Pressure (PSI)0
  • Max Pressure (PSI)400
  • Wetted Materials316 SS
  • Caseglass-reinforced thermoplastic
  • Process Connection1/2\" NPT(M)
  • Unit Of Measurementpsi, kPa, kg/cm², bar
  • Display4-digit LCD, 5/8\" H
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionPressure Gauge 0-400 PSI, 1/2\" NPT(M); 316 SS Sensor/ 316 SS Socket
  • Warranty1 year
1 Year Warranty
Key Features
  • Large display
  • <10 Lux rating- only one foot-candle
  • User selectable functions
  • IP65 weatherproof housing
  • ASME B40.7 conformance
  • 1 year warranty
More About this Item
Threre's no need to ever change a battery or push an on button with these solar-powered digital industrial gauges. The special features of this gauge make it an incredible low-cost solution for many industrial applications. The gauge can run on very low light–<10 Lux rating–that's only one foot-candle!