Weller 037103059963 40-Watt Basic Soldering Station; 120 VAC
Representative image only.
Weller Mfr # WLC100 – Item # EW-97105-63
Designed for applications that need more control than a solder gun produces
$80.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Power (VAC)120
  • Description40-Watt Basic Soldering Station; 120 VAC
Includes
power unit, soldering pen, screwdriver tip, stand and sponge
More About this Item
This soldering station has rapid temperature heat-up and recovery, with variable power control between 5 and 40 watts. The unit also features an on/off switch with indicator, safety holder for the soldering pen, and a iron-plated copper soldering tip. The ergonomically designed pen is slim and comfortable which reduces operator fatigue during extended use.