Weller 037103475428 Professional Soldering Kit, 120V, 260/200 Watts
Representative image only.
Weller Mfr # D550PK – Item # EW-97105-62
Bring the fix to the application with this reliable heavy-duty soldering kit
SPECS & DESCRIPTION
Specifications & Description
  • Set IncludesSoldering gun, 1 soldering tip, 1 smoothing tip, 1 cutting tip, flux brush, soldering aid tool, coil of lead-free solder
  • DescriptionProfessional Soldering Kit, 120V, 260/200 Watts
More About this Item
This professional soldering gun is designed for everyday use and comes in a plastic case with the most common used accessories. The soldering gun has a high/low heat setting, by pulling the trigger into position 1 the gun will produce 260 watts, in position 2 it will produce 200 watts. Other features include a 6 second quick start, high efficiency soldering tips, and two lights designed to illuminate the work surface.