Weller ETA Soldering pencil tip, Screwdriver, 0.062"
Representative image only.
Weller Mfr # ETA – Item # EW-97110-54
ESD safe to protect sensitive components
  • Designed for continuous production soldering
PRODUCT OPTIONS
$11.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSoldering pencil tip, Screwdriver, 0.062"
Key Features
  • Designed for continuous production soldering
More About this Item
These soldering stations feature rapid temperature heat-up and recovery. Wireless temperature lockout prevents operators from raising temperatures higher than specified for applications. Reset station temperature readings to match tip sizes and styles with tip offset function. Slim, comfortable pen reduces operator fatigue during extended use. Station automatically powers down after 90 minutes of inactivity.
Model 97110-00 features an analog temperature dial.
Model 97110-02 features a digital LED for displaying tip temperatures and features a °F/C switch.
What’s included: power unit, soldering pen, screwdriver tip, stand and sponge.
Call Us At1-847-549-7600
Live chat