Whatman 10461000 CYTIVA Whatman™ Polysulfone Filter Holder for 25-mm membranes
Representative image only.
Whatman Mfr # 10461000 – Item # EW-29822-02
In-line removal of particles or microbial contaminants
  • Holds 25-mm membrane
  • Max temperature 250°F (121°C)
  • Max pressure 100 psi
  • Autoclavable
$226.75USD / Pkg of 10
SPECS & DESCRIPTION
Specifications & Description
  • Filter Diameter (mm)25
  • Body MaterialPolysulfone
  • DescriptionPolysulfone Filter Holder for 25-mm membranes
Key Features
  • Holds 25-mm membrane
  • Max temperature 250°F (121°C)
  • Max pressure 100 psi
  • Autoclavable
More About this Item
Use this polysulfone holder with silicone O-ring for in-line removal of particles or microbial contaminants from liquids or gases, as in liquids or cold sterilization. Removable tubing connectors on both ends fit 3/8" ID tubing. Includes luer-lock fitting to permit use with syringe. Use holder as an aseptic bell.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.