Whatman 1540-150 540 Ashless Quantitative Filter Paper, 8um, 15cm; 100/Box
Representative image only.
Whatman Mfr # 1540-150 – Item # EW-06647-45
The industry standard for high-purity filtration
  • Available in a variety of grades and sizes
  • 45 day warranty
PRODUCT OPTIONS
$130.50USD / Box of 100
SPECS & DESCRIPTION
Specifications & Description
  • MaterialCotton Fiber
  • Pore Size (micron)8
  • Diameter (mm)150
  • Diameter (cm)15
  • Description540 Ashless Quantitative Filter Paper, 8um, 15cm; 100/Box
  • Warranty45 days
Key Features
  • Available in a variety of grades and sizes
  • 45 day warranty
More About this Item
Our CYTIVA Whatman™ filter papers offer you flexibility in filter choice without sacrificing quality. All filters are packaged 100 per box.
Quantitative filter papers are the choice for gravimetric analysis. High-purity hardened filters have a high wet-strength and are chemical-resistant to handle vacuum filtration or acid/alkali solutions.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.