Whatman 1841-090 Glass Microfiber Prefilter, 1um, 90 mm; 20/Pk

Representative image only.
Whatman Mfr # 1841-090 – Item # EW-29829-24
For 500- and 1000-mL units
  • 1.0 µm pore
  • 90-mm diameter
  • Pack of 20

$71.50USD / Pkg of 20

SPECS & DESCRIPTION

Specifications & Description

  • Membrane MaterialGlass Microfiber (GMF)
  • Pore Size (µm)1
  • Membrane Diameter (mm)90
  • DescriptionGlass Microfiber Prefilter, 1um, 90 mm; 20/Pk

Key Features

  • 1.0 µm pore
  • 90-mm diameter
  • Pack of 20

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat