Whatman 6714-6004 CYTIVA Whatman™ Groundwater Filter Capsule, 0.45 µm, 1/cs
Representative image only.
Whatman Mfr # 6714-6004 – Item # EW-29705-92
Filter ground water samples for dissolved metal analysis
  • Suitable for use in EPA Method 3005
  • 45 day warranty
$87.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolyethersulfone
  • Pore Size (micron)0.45
  • Housing MaterialPolypropylene
  • Connections1/4\" to 3/8\" hose barb
  • Effective Filtration Area (cm²)600
  • Surface Area (m²)0.06
  • Max Pressure (PSI)60
  • Max Pressure4.1
  • DescriptionGroundwater Filter Capsule, 0.45 µm, 1/cs
  • Warranty45 days
EPA United States Environmental protection agency
Key Features
  • Suitable for use in EPA Method 3005
  • 45 day warranty
More About this Item
Whatman groundwater filter capsule is designed to make the process of collecting ground water samples for analysis quick, easy, and convenient. It is the perfect choice for anyone who must meet strict environmental protocol standards. Capsules contain no glues, adhesives, metals, epoxies or other extraneous materials. Filter housing is made of polypropylene and all housing components are thermally fused. Capsules provide at least 600 cm2 of effective filtration area. Connections are 1/4 to 3/8" (6-9 mm) stepped barb (SB). Max operating pressure is 60 psi (4.1 bar).
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.