Whatman G2 Mini-UniPrep Filters, Clr, 0.45um, Glass MF, SC; 1000/Pk
Representative image only.
Whatman Mfr # GN503NPUGMF – Item # EW-32818-57
Prepare samples three times faster than traditional syringe filtration methods
  • Decrease sample preparation time and sample loss due to transferring containers
  • All-in-one filtration system replaces syringe filter, syringe, glass vial, cap, and septa
  • New glass design minimizes leachables and filters quicker than traditional syringe filtration methods
  • Designed to load directly into your autosampler while minimizing instrument downtime from unfiltered samples
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionMini-UniPrep Filters, Clr, 0.45um, Glass MF, SC; 1000/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Decrease sample preparation time and sample loss due to transferring containers
  • All-in-one filtration system replaces syringe filter, syringe, glass vial, cap, and septa
  • New glass design minimizes leachables and filters quicker than traditional syringe filtration methods
  • Designed to load directly into your autosampler while minimizing instrument downtime from unfiltered samples
  • 1 year warranty
More About this Item
These borosilicate glass autosampler vials with integrated plunger, filter membrane, and septum cap make UHPLC/HPLC sample filtration easy! Simply load 220 to 500 µL of your sample into the vial, place the plunger into the neck of the vial, and place the assembled filter into the hand compression device. The sample will only come into contact with plastic momentarily as the plunger is depressed. Once compressed, the Mini-UniPrep G2 syringeless filters are equivalent to a 12 x 32 mm autosampler vial—perfect for loading directly into your UHPLC/HPLC instrument.
Capacity:
Unfiltered sample chamber: 500 µL
Filtered sample chamber: 330 µL
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.
Minimum filtering sample volume: 220 µL placed into the chamber to obtain a 50 µL filtered sample
Materials of construction: borosilicate glass chamber, PP plunger outer housing, borosilicate glass plunger inner storage vial, silicone septa with PTFE liner, PP cap
Maximum operating temperature: 122°F (50°C)
Available for sale in the USA only