Whatman In-Line Solvent Degasser, Aqueous Phase; 10/box
Representative image only.
Whatman Mfr # 6726-5002 – Item # EW-29702-00
Simultaneously filter and degas solvents to provide better HPLC reproducibility
  • Eliminate false peaks due to solvent impurities and gas bubbles
PRODUCT OPTIONS
$270.50USD / Box of 10
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionIn-Line Solvent Degasser, Aqueous Phase; 10/box
Key Features
  • Eliminate false peaks due to solvent impurities and gas bubbles
More About this Item
Degassers simultaneously filter and degas solvents to provide better HPLC reproducibility. They also eliminate false peaks due to solvent impurities and gas bubbles. Degassers feature built-in 0.2-µm filter. Aqueous phase degasser has a nylon filter and organic phase degasser has a polypropylene filter. Other wetted parts are polypropylene. Each box of degassers includes one set of ferrule nuts for 1/8" OD tubing.
CYTIVA Whatman™ Warranties to served market customers are in lieu of all other warranties, expressed or implied, and CYTIVA Whatman™ disclaims all other warranties, including without limitation, the implied warranties of non-infringement, merchantability, fitness for a particular purpose, quiet enjoyment, system integration and data accuracy. CYTIVA Whatman™ limited warranties set forth in the CYTIVA Whatman™ warranty statement shall be enforced by and only by the served market customers.