Wheaton 776580 Rubber bottle leash, pack of 25 (BOD bottles)
Representative image only.
Wheaton Mfr # 776580 – Item # EW-03660-12
Enhance the versitility of your Wheaton BOD Bottles
PRODUCT OPTIONS
$38.50USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRubber
  • Use WithBOD bottles
  • DescriptionRubber bottle leash, pack of 25 (BOD bottles)
More About this Item
Choose from a wide range of BOD bottle accessories.
Model 03660-09 cap and insert is for use with 03660-15 BOD bottles.
The model 03660-12 rubber bottle leash holds the stopper to the bottle to avoid misplacing the stopper or breakage.