Whirl-Pak B01591WA Hydrated PolySponge™ Sampling Bag with Sampling Sponge and Gloves, 18 oz, Sterile; 100/Bx

Representative image only.
Whirl-Pak Mfr # B01591 – Item # EW-00649-92
Make environmental surface sampling faster and easier
  • Write-on label makes identifying samples easy and convenient
  • Each bag contains a polyurethane sponge, premoistened with HiCap™ neutralizing broth
  • Terminally gamma processed to guarantee sterility
  • -Each box contains five foil pouches with 20 sponges each for a total of
  • 100 units. Bags are 9" L x 4" W (23 x 11.5 cm) with a thickness of
  • 2.5 mils (.064 mm).

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (oz)18
  • MaterialPolyurethane
  • Length (in)9
  • Width (in)4
  • Length (cm)22.86
  • Width (cm)10.16
  • Thickness (mil)2.5
  • DescriptionSampling Bag with Sampling Sponge and Gloves, 18 oz, Sterile; 100/Bx
Sterile

Key Features

  • Write-on label makes identifying samples easy and convenient
  • Each bag contains a polyurethane sponge, premoistened with HiCap™ neutralizing broth
  • Terminally gamma processed to guarantee sterility
  • -Each box contains five foil pouches with 20 sponges each for a total of
  • 100 units. Bags are 9" L x 4" W (23 x 11.5 cm) with a thickness of
  • 2.5 mils (.064 mm).

More About this Item

Collection broths play a critical role in the success of an environmental monitoring program. HiCap™ neutralizing broth is the first collection broth developed specifically for use in the food and beverage industry as an alternative to traditional collection broths. The polyurethane sponges included with each bag are nontoxic and do not contain biocides. Unlike the “wet” cellulose manufacturing process, there is no need to include biocides to control microbial growth during the polymerization process.

Model 00649-92 includes a glove.

Durable high-density polyurethane sponge is packed in a sealed
18-oz. (532 ml) write-on Whirl-Pak® bag and hydrated with 16 ml of
HiCap™ Neutralizing Broth.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat