Wide-Range Relief Valve, PVC, 1/4" NPT(F)
Representative image only.
Item # EW-01350-60
Prevent leaks with the emergency shutdown design
  • Three-seal system vents liquid to warn of approaching maintenance
PRODUCT OPTIONS
$269.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port1/4" NPT(F)
  • Connection TypeNPT(F)
  • Inlet Port1/4" NPT(F)
  • Max Pressure (PSI)210
  • Wetted MaterialsPVC and Viton
  • Media CompatibilityLiquids
  • DescriptionWide-Range PVC Relief Valve, 1/4" NPT(F)
Key Features
  • Three-seal system vents liquid to warn of approaching maintenance
More About this Item
Valves with connections of 1/4" or 1/2" have a diaphragm made of Viton® and withstand a maximum inlet pressure of 210 psig. All other valves have a PTFE shaft design and withstand a maximum inlet pressure of 140 psig. The shaft design prevents emergency shutdown with a three-seal system that will vent liquid to warn of approaching maintenance. All valves are recommended for liquid applications.