Wika D010300WI Ti.D01 Adjustable Angle Solar Thermometer, 3.5" L without Well; -50/300F
Representative image only.
Wika Mfr # D010300WI – Item # EW-90552-49
$82.00USD / Each
sign in for your price.
$75.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)-50
  • Max Temperature (° F)300
  • Dial / Case Material304 Stainless Steel
  • Stem Length (in)3.5
  • Stem Material304 Stainless Steel
  • Connection0.75" NPT(M)
  • DescriptionTi.D01 Adjustable Angle Solar Thermometer, 3.5" L without Well; -50/300F
More About this Item
The Solar Industrial thermometer eliminates mercury filled glass thermometers and offers users fast, accurate, and easy-to-read temperature indications. This thermometer features a totally adjustable case to permit viewing at any angle, and its bulb and socket are completely interchangeable with standard industrial glass thermometers.
The Solar Industrial thermometer is switchable between Fahrenheit and Celsius, and offers a sensing range of -50 to 300degF and -50 to 150degC, resolved in tenths of a degree, with accuracy to within 1% of reading. The solar panels on the instrument require only 10 LUX (one foot-candle) of light to power the 3/8" LCD digits, and the thermometer updates its reading every 10 seconds. The case is made of high-impact ABS, and the thermometer is availa