Wika IS-20 Intrinsically Safe Compound Transmitter, 30" Hg to 30 psig

Representative image only.
Wika Mfr # 12127833 – Item # EW-68446-05
  • FEATURES
  • Pressure ranges from 50 INWC to 15,000 PSI
  • FM, CSA approval for:
  • Intrinsically safe Class I, II and Division 1, Group A, B, C, D, E, F, G
  • Dust Class II and III Division 1, Group E, F, G
  • Class I, Zone 0, AEx ia II C
  • Ex-protection EEx ia I/II C T6 accordin to ATEX for
  • Gases, vapors and mist: Connection to Zone 0, Zone 1, Zone 2
  • Dust: Connection to Zone 20, Zone 21, Zone 22
  • Mining: Category M1 and M2
  • 2 year warranty
  • SPECS & DESCRIPTION

    Specifications & Description

    • Accuracy±0.25% BFSL
    • Max Pressure (PSI)30
    • Min Temperature (° F)-4
    • Max Temperature (° F)176
    • Min Temperature (° C)-20
    • Max Temperature (° C)80
    • Max Vacuum (\" Hg)30
    • Wetted Materials316L SS
    • Process Connection1/2\" NPT(M)
    • Battery Type10 to 30 VDC
    • Electrical Connections1/2\" NPT(F) conduit
    • Electrical terminationScrew terminals
    • Output4 to 20 mA
    • Housing316L SS
    • Media CompatibilityGases and liquids compatible with wetted materials
    • DescriptionIntrinsically Safe Compound Transmitter, 30\" Hg to 30 psig
    • Warranty2 year
    Two year warranty

    Key Features

    • FEATURES
    • Pressure ranges from 50 INWC to 15,000 PSI
    • FM, CSA approval for:
    • Intrinsically safe Class I, II and Division 1, Group A, B, C, D, E, F, G
    • Dust Class II and III Division 1, Group E, F, G
    • Class I, Zone 0, AEx ia II C
  • Ex-protection EEx ia I/II C T6 accordin to ATEX for
  • Gases, vapors and mist: Connection to Zone 0, Zone 1, Zone 2
  • Dust: Connection to Zone 20, Zone 21, Zone 22
  • Mining: Category M1 and M2
  • 2 year warranty
  • More About this Item

    The IS-20 series of intrinsically safe pressure
    transmitters are designed for industrial pressure
    measurement applications in hazardous
    areas where intrinsically safe ratings are required.
    The stainless steel wetted parts feature an
    all-welded measuring cell for improved media
    compatibility. There are no internal soft sealing
    materials that may react with the media or
    deteriorate over time.
    The IS-21-S and IS-21-F transmitters feature
    a flush diaphragm process connection. They are
    specifically designed for the measurement of
    viscous fluids or media containing solids that
    may clog a NPT process connection.
    Models IS-20-F and IS-21-F feature an integral
    stainless steel junction box with internal terminal
    block for use in extremely harsh environments.
    A 1/2'' NPT female conduit connection is standard
    on all models and a cable compression electrical
    connection is available as an option.
    All models require a 10 to 30 volt supply provided
    by an intrinsically safe power supply or
    through an approved intrinsically safe zener
    diode barrier.

    REVIEWS

    Q&A

    CUSTOMER SUPPORT

    Need Help with this product?

    Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

    Call Us At1-800-323-4340
    Live chat