Wildco 1728-G30 grab dredge, 6" x 6" sampling area
Representative image only.
Wildco Mfr # 1728-G30 – Item # EW-05471-10
Quickly sample sediment from various bodies of water
  • Dredges are constructed of corrosion-resistant stainless steel
  • Jaw weights and arms are zinc-plated
  • Top surfaces are covered with #30-mesh screen to reduce shock waves and drifts, and to prevent sediments and organisms from escaping
  • 3 month warranty
$1,445.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialStainless Steel
  • Descriptiongrab dredge, 6\" x 6\" sampling area
  • Warranty3 month
3 Month Warranty
Key Features
  • Dredges are constructed of corrosion-resistant stainless steel
  • Jaw weights and arms are zinc-plated
  • Top surfaces are covered with #30-mesh screen to reduce shock waves and drifts, and to prevent sediments and organisms from escaping
  • 3 month warranty
More About this Item
Designed to sample all types of benthos sediments (except the hardest clay) in both fresh and salt water. Closing mechanism releases when dredge strikes bottom; locking “safety pin” prevents accidental closing.