Worthington D025-8C00 Storage Dewars Tipping Stand
Representative image only.
Worthington Mfr # D025-8C00 – Item # EW-03773-70
$1,540.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionStorage Dewars Tipping Stand
More About this Item
Easily move storage Dewars and Dewars with canister storage systems. Sturdy cast aluminum frame has five wheels with ball-bearing swivel casters. Use model 03773-65 for Dewars with 15.6" diameter base; use 03773-71 for Dewars with 18.8" diameter base. Shpg wt 8.0 lb (3.7 kg). Note: Roller base does not include tipping stand.
Tipping stand supports the weight of the Dewar
while you safely tip the unit. For model 03773-57 only