WTW 201570 DurOx 325-3 Replacement DO Probe
Representative image only.
WTW Mfr # 201570. – Item # EW-53105-43
Stable signal thanks to special membrane technology
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter TypeAccessory; Probes and Sensors
  • Description201570 DurOx 325-3 Replacement DO Probe
  • Warranty3 year
3 Year WarrantyISO9001:2000 Certified Supplier
Key Features
  • 3 year warranty
More About this Item
Robust, membrane-covered galvanic dissolved oxygen sensor for applications in flowing waters, fish hatcheries, aeration basins and more fields. With screw-on protection, the special membranes achieve stable measured values, especially in the aeration basin and similar applications with strongly fluctuating oxygen content.
This probes features measuring ranges of 0 to 50 mg/l O2 concentration, 0 to 600% O2 saturation, and 0 to 1250 mbar O2 partial pressure. This probes offers a temperature range of 0 to 50°C.