YSI 3140 platinizing solution for use with platinizing station
Representative image only.
YSI Mfr # 86511 – Item # EW-01486-20
  • 3140 platinizing solution; replacement
PRODUCT OPTIONS
$245.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Use WithPlatinizing station
  • Descriptionplatinizing solution for use with platinizing station
Key Features
  • 3140 platinizing solution; replacement
More About this Item
Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.
REVIEWS
REVIEW SNAPSHOT®
by PowerReviews
4.0
(based on 1 review)
Reviewed by 1 customer
Displaying review 1
Back to top
4.0
just plain works
By 
from canada
Pros
  • Works as advertised
Cons
  • Expensive
Best Uses
  • Pt Black Deposition
Worked well from the first time I used it. Nice even Pt coat in 2 minutes.
  • Was this a gift?:
  • No
Displaying review 1
Back to top