YSI 3403 Model Conductivity Cell, Glass, Microcell, K=1
Representative image only.
YSI Mfr # 603403 – Item # EW-01485-30
Optimize the performance of YSI meters
  • Choose from a variety of accessories
  • 3403 conductivity probe, glass, microcell
  • 1 year warranty
PRODUCT OPTIONS
$749.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialGlass
  • Compatible WithYSI 3100 or 3200
  • Length (mm)178
  • Diameter (mm)13
  • DescriptionModel Conductivity Cell, Glass, Microcell, K=1
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from a variety of accessories
  • 3403 conductivity probe, glass, microcell
  • 1 year warranty
More About this Item
Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.