YSI 3417 Model Conductivity cell, ABS Plastic, K=1
Representative image only.
YSI Mfr # 099820 – Item # EW-01485-40
Optimize the performance of YSI meters
  • Choose from a variety of accessories
  • 3417 conductivity cell
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialABS plastic
  • Compatible WithYSI 3100 or 3201
  • Length (mm)146
  • Diameter (mm)13
  • DescriptionModel Conductivity cell, ABS Plastic, K=1
  • Warranty1 year
1 Year Warranty
Key Features
  • Choose from a variety of accessories
  • 3417 conductivity cell
  • 1 year warranty
More About this Item
Replacement electrodes, cables, power adapters, and other accessories for your YSI water analysis meters.