YSI ODO/T Optical Dissolved Oxygen Probe and Cable Assembly, 100 m Cable
Representative image only.
YSI Mfr # 627200-100 – Item # EW-59353-77
Get lab-quality results at the source
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Smart probes store calibration information for up to one year saving you time and money
  • Includes 100-m cable
PRODUCT OPTIONS
$1,020.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • ApplicationsWater quality, waste water, natural resources, oxygen analysis
  • Min Temperature (° C)-5
  • Max Temperature (° C)70
  • Temperature CompensationYes
  • GuardStainless steel
  • ConnectorWaterproof
  • Cable Length (m)100
  • DescriptionOptical Dissolved Oxygen Probe and Cable Assembly, 100 m Cable
Two year warranty
Key Features
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Smart probes store calibration information for up to one year saving you time and money
  • Includes 100-m cable
More About this Item
Interchangeable digital "smart" probes store calibration data for a year, saving you time and calibration solution costs—no need to recalibrate if connected to different meters. Convenient cable management kits are included with probes featuring cable lengths from 13-ft (4-m) to 100-ft (30-m)