This item is now discontinued. View the direct replacement: EW-35613-28 or browse similar items

YSI PH100 Ecosense pH100A pH/mV/Temp Handheld Meter Only

Representative image only.
YSI Mfr # 606075 – Item # EW-59351-32
Accurate, economical portable meter
  • Automatic buffer recognition for US and NIST buffers
  • Automatic or manual temperature compensation

SPECS & DESCRIPTION

Specifications & Description

  • Meter TypePortable
  • Display typeLCD
  • Min pH (pH)-2
  • Max pH (pH)16
  • pH Resolution0.01/0.1
  • pH Accuracy±0.1 pH
  • Buffer RecognitionNIST and USA
  • CalibrationUp to 2 points
  • Min mV (mV)-1999
  • Max mV (mV)1250
  • mV Resolution1 or 10 mV
  • mV Accuracy±0.1% + 1 digit
  • Min Temperature (° C)-10
  • Max Temperature (° C)110
  • Min Temperature (° F)14
  • Max Temperature (° F)230
  • Temperature compensationAutomatic or manual
  • WaterproofYes
  • Battery1 x 9 V battery (included)
  • Width (cm)18.415
  • Height (cm)7.9375
  • Width (in)7 1/4
  • Length (in)1 1/2
  • Height (in)3
  • DescriptionEcosense pH100A pH/mV/Temp Handheld Meter Only

Key Features

  • Automatic buffer recognition for US and NIST buffers
  • Automatic or manual temperature compensation

More About this Item

The pH100A combines low cost with a rugged, waterproof design--making it ideal for spot sampling in wastewater, surface water, or aquaculture. Meter features easy-to-use interface, one-hand operation, automatic or manual temperature compensation, electrode offset recognition, and electrode efficieny display. Meter also has low battery indicator, auto shutoff, and nonvolatile 50 data sets memory with calibration data. Calibrate the meter quickly using the internal buffer recognition feature.

Purchase meter with probes separately to customize your system, or choose a kit for a complete system.

Order electrode separately.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat