YSI SI Analytics Micro Double Platinum Electrode for KF Titrators
Representative image only.
YSI Mfr # KF 1100 – Item # EW-24907-55
Keep titration running smoothly
  • Connection cables
  • Replacement electrodes
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithYSI Karl Fischer Titrators
  • DescriptionMicro Double Platinum Electrode for YSI KF Titrators
Key Features
  • Connection cables
  • Replacement electrodes
More About this Item
Convenient parts and accessories to keep your YSI titrator operating at its best.
Micro-double platinum combination electrode, for Karl-Fischer titrations, with fixed cable (1-m), double platinum pin and tapper NS 7.5 for TZ 1770 and TZ 1772.