Zippette 91003 Dispensor BT 0-10.0mL

Representative image only.
Zippette Mfr # 91003 – Item # EW-94702-70
Simple and durable — ideal for use in the lab, the field, or the classroom
  • Available in five fully adjustable volume ranges
  • Accuracy of delivery to within ± 0.3% on maximum delivery
  • Fully autoclavable at 121°C, 2 bar
  • All models are fitted with a new anti-drip safety value that guards against drips when not in use
  • Chemical-resistant liquid pathway allows a wide range of liquids to be safely dispensed

SPECS & DESCRIPTION

Specifications & Description

  • Division (mL)0.05
  • Accuracy±0.3%
  • Reproducibility±0.1%
  • AutoclavableYes
  • Wetted PartsBorosilicate glass and PTFE
  • DescriptionDispensor BT 0-10.0mL

Key Features

  • Available in five fully adjustable volume ranges
  • Accuracy of delivery to within ± 0.3% on maximum delivery
  • Fully autoclavable at 121°C, 2 bar
  • All models are fitted with a new anti-drip safety value that guards against drips when not in use
  • Chemical-resistant liquid pathway allows a wide range of liquids to be safely dispensed

More About this Item

Redesigned twist-to-lock volume adjuster stays secure to ensure accuracy. To dispense, simply set volume, lock in adjuster, then lift piston and push down. A delivery tube of PTFE lets you safely fill handheld narrow-neck containers. Piston assembly is easy to disassemble for cleaning.
All models have a polypropylene-encased body, borosilicate glass syringe, chemical-resistant plunger, and a safety valve system contained within a PVDF pedestal that holds a prime — prevents reagent loss during re-priming. Wetted parts are borosilicate glass and PTFE. Dispensers fit bottles with 30-mm necks and include adapters for other common bottle neck sizes. Order bottles separately.
Model 94702-73 adds digital volume control. Simply turn the top button to activate the digital LCD to set the desired volume. An auto-off feature turns off the digital display after three minutes of non-use to prolong battery life.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat