This product is now discontinued, View Related Products

1,2,3,4-Tetrahydronaphthalene

Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical T724 1,2,3,4-Tetrahydronaphthalene (Laboratory) (4 liter) EW-88087-23 Mfr # T724
COMPARE
119-64-2
Call For Price
Acros Organics AC146730010 1,2,3,4-Tetrahydronaphthalene 98+% (1l) CAS 119-64-2 EW-88145-29 Mfr # AC146730010
COMPARE
119-64-2
Call For Price
Acros Organics AC146730025 1,2,3,4-Tetrahydronaphthalene 98+% (2.5l) EW-88145-30 Mfr # AC146730025
COMPARE
119-64-2
Call For Price
Acros Organics AC187670010 1,2,3,4-Tetrahydronaphthalene 95-98% (1l) EW-88200-62 Mfr # AC187670010
COMPARE
119-64-2
Call For Price
Acros Organics AC187670025 1,2,3,4-Tetrahydronaphthalene 95-98% (2.5l) EW-88200-63 Mfr # AC187670025
COMPARE
119-64-2
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat