This product is now discontinued, View Related Products

1,3-Diaminopropane

Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC112350010 1,3-Diaminopropane 99% (1l) EW-88103-94 Mfr # AC112350010
COMPARE
109-76-2
Call For Price
Acros Organics AC112352500 1,3-Diaminopropane 99% (250 mL) EW-88103-95 Mfr # AC112352500
COMPARE
109-76-2
Call For Price
Acros Organics AC219801000 1,3-Diaminopropane 99+% (100 mL) EW-88235-12 Mfr # AC219801000
COMPARE
109-76-2
Call For Price
Acros Organics AC219805000 1,3-Diaminopropane 99+% (500 mL) EW-88235-13 Mfr # AC219805000
COMPARE
109-76-2
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat