This product is now discontinued, View Related Products

Fisher Chemical 1-Hexanesulfonic Acid Sodium Salt

Quality and consistency your lab can count on
  • Assay (by Sulfated Sodium Ash) - ≥98%
  • Solubility (1g/25mL H2O) - To pass test
  • Optical Absorbance at 220nm - ≤0.1
  • at 254nm - ≤0.015
  • at 300nm - ≤0.010
  • Ultraviolet Absorbance Curves are measured using a 1g/100mL solution run
  • against distilled H2O set at zero absorbance.
Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (g)
Availability Pricing
Fisher Chemical O307225 1-Hexanesulfonic Acid Sodium Salt (HPLC) (25g) EW-88077-81 Mfr # O307225
COMPARE
2832-45-3 HPLC 25
Call For Price
Acros Organics AC411600050 1-Hexanesulfonic acid sodium salt, HPLC grade (5g) EW-88348-64 Mfr # AC411600050
COMPARE
2832-45-3
Call For Price
Acros Organics AC411600250 1-Hexanesulfonic acid sodium salt, HPLC grade (25g) EW-88348-65 Mfr # AC411600250
COMPARE
2832-45-3 25
Call For Price
Acros Organics AC411601000 1-Hexanesulfonic acid sodium salt, HPLC grade (100g) EW-88348-66 Mfr # AC411601000
COMPARE
2832-45-3
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat