This product is now discontinued, View Related Products

2,3-Butanedione monoxime 29152

Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC150371000 2,3-Butanedione monoxime 99+% (100g) EW-88151-47 Mfr # AC150371000
COMPARE
57-71-6
Call For Price
Acros Organics AC150375000 2,3-Butanedione monoxime 99+% (500g) EW-88151-48 Mfr # AC150375000
COMPARE
57-71-6
Call For Price
Acros Organics AC403300250 2,3-Butanedione monoxime 96% (25g) EW-88340-96 Mfr # AC403300250
COMPARE
57-71-6
Call For Price
Acros Organics AC403301000 2,3-Butanedione monoxime 96% (100g) EW-88340-97 Mfr # AC403301000
COMPARE
57-71-6
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat