This product is now discontinued, View Related Products

Acros Organics 2,6-Dichlorophenol, 99%

Assay 99%
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC113590010 2,6-Dichlorophenol 99% (1kg) EW-88105-98 Mfr # AC113590010
COMPARE
87-65-0
Call For Price
Acros Organics AC113590250 2,6-Dichlorophenol 99% (25g) EW-88105-99 Mfr # AC113590250
COMPARE
87-65-0
Call For Price
Acros Organics AC113591000 2,6-Dichlorophenol 99% (100g) EW-88106-00 Mfr # AC113591000
COMPARE
87-65-0
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat