This product is now discontinued, View Related Products

Acros Organics 2-Pentanone, 98+%

Reagents and solutions to meet your lab and research needs
  • Quality and consistency your lab can count on
Quality and consistency your lab can count on.
Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Fisher Chemical BP2636250 2-Pentanone (&gt2;98%) (250mL) EW-88061-26 Mfr # BP2636250
COMPARE
107-87-9
Call For Price
Acros Organics 2-Pentanone, 98+%, Liquid; 250 mL EW-88122-28 Mfr # AC123502500
COMPARE
107-87-9
Call For Price
Acros Organics AC130000010 2-Pentanone 97% (1l) 8813185 EW-88131-85 Mfr # AC130000010
COMPARE
107-87-9
Call For Price
Acros Organics AC130000050 2-Pentanone 97% (5ml) EW-88131-86 Mfr # AC130000050
COMPARE
107-87-9
Call For Price
Acros Organics AC130005000 2-Pentanone 97% (500ml) EW-88131-87 Mfr # AC130005000
COMPARE
107-87-9
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat