This product is now discontinued, View Related Products

3,5-Dimethylphenol 30930

Call For Price

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
CAS Number Purity Grade Weight (mL)
Availability Pricing
Acros Organics AC116600010 3,5-Dimethylphenol 98% (1kg) EW-88110-88 Mfr # AC116600010
COMPARE
108-68-9
Call For Price
Acros Organics AC116600050 3,5-Dimethylphenol 98% (5g) EW-88110-89 Mfr # AC116600050
COMPARE
108-68-9
Call For Price
Acros Organics AC116602500 3,5-Dimethylphenol 98% (250g) EW-88110-90 Mfr # AC116602500
COMPARE
108-68-9
Call For Price
Acros Organics AC162791000 3,5-Dimethylphenol 99+% (100g) EW-88171-08 Mfr # AC162791000
COMPARE
108-68-9
Call For Price
Acros Organics AC162795000 3,5-Dimethylphenol 99+% (500g) EW-88171-09 Mfr # AC162795000
COMPARE
108-68-9
Call For Price
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat