304 SS Submersible Moderate-Flow Dewatering Pumps
Semi-open impeller design pumps solids up to 3/8" in slurries and up to 81 GPM
  • 304 stainless steel construction resists corrosion and rust
  • Pumps with automatic operation have float switch; manual operation models activate with a switch
  • Air-filled motors have permanent split capacitor design with built-in thermal protection and automatic reset
Operate pumps either fully or partially submerged. Pumps are ideal for slurries and dirty or semi-dirty waters.
$399.60 - $1,544.50USD / Each
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPM) Power (VAC) Motor hp
Availability Pricing
COMPARE
45 115 1/3
$418.20USD / Each
COMPARE
45 115 1/3
$439.20USD / Each
COMPARE
45 115 1/3
$399.60USD / Each
COMPARE
42 115 1/3
$644.25USD / Each
COMPARE
42 115 1/3
$623.25USD / Each
COMPARE
68 115 1/2
$1,229.50USD / Each
COMPARE
68 115 1/2
$1,219.00USD / Each
COMPARE
68 230 1/2
$1,156.00USD / Each
COMPARE
68 460 1/2
$1,156.00USD / Each
COMPARE
68 115 3/4
$1,460.50USD / Each
COMPARE
68 115 3/4
$1,418.50USD / Each
COMPARE
68 230 3/4
$1,345.00USD / Each
COMPARE
68 460 3/4
$1,261.00USD / Each
COMPARE
80 230 1
$1,460.50USD / Each
COMPARE
80 460 1
$1,450.00USD / Each
COMPARE
80 230 1
$1,544.50USD / Each
COMPARE
80 460 1
$1,534.00USD / Each
Showing 1- 17 of 17