This product is now discontinued, View Related Products
3M Three-Panel Particulate Respirator
Lightweight respirator delivers comfort, convenience, and security
  • Improves visibility, prevents collapsing, and fits many face sizes
  • Individually wrapped to prevent contamination during storage
Low profile makes these respirators ideal for use with other safety equipment. The soft inner panel feels comfortable to the skin. The adjustable nose clip provides a custom fit and a long-lasting, secure seal.
Use in textile, dry chemical, surface preparation, food preparation, woodworking, milling, mineral processing, and other industries that create dusty environments. Respirator can also be used in environments with liquid or non-oil based sprays that do not contain harmful vapors.
Approval: NIOSH
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Rating Description
Availability Pricing
3M 9210 Three-Panel Particulate respirator, N95, Box of 20 EW-86401-00 Mfr # 9210
COMPARE
N95 Three-Panel Particulate respirator, N95, Box of 20
Call For Price
3M 9211 Three-Panel Particulate respirator with exhalation valve, N95, Box of 10 EW-86401-10 Mfr # 9211
COMPARE
N95 Three-Panel Particulate respirator with exhalation valve, N95, Box of 10