Thermo Scientific™ National™ Vial Kits
Save time during sample preparation and reduce the risk of contamination
  • Choose from glass or polypropylene vials
4-mL Vials for 48-position autosamplers measure 15 x 45 mm. All vials are offered in convenience packs that include 100 vials and 100 cap/septum assemblies.
$58.70 - $150.75USD / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Septum Type Tube Material Volume (mL)
Availability Pricing
Thermo Scientific Vial Kit, 4 mL, Polypropylene, Polyethylene  Plug; 100/Pk EW-98814-30 Mfr # C4015-47P
COMPARE
Built in cover Polypropylene 4
$58.70USD / Pkg of 100
Thermo Scientific Vial Kit, 4 mL, Clear Glass, Polypropylene Cap with PTFE/Silicone Septa; 100/Pk EW-98814-90 Mfr # C4015-88
COMPARE
Red PTFE/white silicone Borosilicate Glass 4
$150.75USD / Pkg of 100