4 to 20 mA Pace Pumps
The 4 to 20 mA pumps feature automatic control with 4 to 20 mADC direct or external pacing, reliable timing circuits, stop function, and manually adjustable stroke rate and length. Operate pump in automatic or manual mode by selecting the proper setting on the "Auto-Off-Manual" switch. Circuit protection guards against voltage and current upsets. Install pumps indoors or outdoors.
$965.00 - $2,328.00USD / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (mL/min) Max Pressure (PSI) Power (VAC)
Availability Pricing
Pulsafeeder LPA2MA-VTC1-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 6 GPD, 115 VAC EW-74141-00 Mfr # LPA2MA-VTC1-XXX
COMPARE
16 150 115
$965.00USD / Each
Pulsafeeder LPA2MB-VTC1-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 6 GPD, 230 VAC EW-74141-05 Mfr # LPA2MB-VTC1-XXX
COMPARE
16 150 230
$1,019.00USD / Each
Pulsafeeder LPB3MA-VTC1-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 12 GPD, 115 VAC EW-74141-10 Mfr # LPB3MA-VTC1-XXX
COMPARE
32 150 115
$1,036.00USD / Each
Pulsafeeder LPD4MA-VTC1-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 21 GPD, 115 VAC EW-74141-20 Mfr # LPD4MA-VTC1-XXX
COMPARE
55 150 115
$1,153.00USD / Each
Pulsafeeder LPG4MA-VTC1-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 42 GPD, 115 VAC EW-74141-30 Mfr # LPG4MA-VTC1-XXX
COMPARE
110 150 115
$1,530.00USD / Each
Pulsafeeder LPG4MB-VTC1-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 42 GPD, 230 VAC EW-74141-35 Mfr # LPG4MB-VTC1-XXX
COMPARE
110 150 230
$1,552.00USD / Each
Pulsafeeder LPG5MA-VTC3-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 94 GPD, 115 VAC EW-74141-40 Mfr # LPG5MA-VTC3-XXX
COMPARE
247 100 115
$1,515.00USD / Each
Pulsafeeder LPG5MB-VTC3-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 94 GPD, 230 VAC EW-74141-45 Mfr # LPG5MB-VTC3-XXX
COMPARE
247 100 230
$1,549.00USD / Each
Pulsafeeder LPH7MA-WTC3-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 240 GPD, 115 VAC EW-74141-50 Mfr # LPH7MA-WTC3-XXX
COMPARE
631 35 115
$1,685.00USD / Each
Pulsafeeder LPH8MA-WTCB-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 500 GPD, 115 VAC EW-74141-60 Mfr # LPH8MA-WTCB-XXX
COMPARE
1314 20 115
$2,272.00USD / Each
Pulsafeeder LPH8MB-WTCB-XXX E Plus Solenoid Diaphragm Pump, 4 to 20 mA, 500 GPD, 230 VAC EW-74141-65 Mfr # LPH8MB-WTCB-XXX
COMPARE
1314 20 230
$2,328.00USD / Each
Showing 1- 11 of 11