4-Watt, Short/Long-Wave Ultraviolet Lamp
Get short or longwave UV light at the touch of a button.
Compact, short/long-wave lamp comes with two 4-watt tubes (254 and 365 nm). Each tube produces a distinct wavelength; no mixing of primary UV ranges occurs. Order optional lamp stand 09817-00 separately.
$557.00 - $568.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Power (Hz) Power (VAC) Wavelength
Availability Pricing
Analytik Jena Short/long-wave UV lamp; 4 watts, 254/365 nm wavelength, 115 VAC/60 Hz EW-09817-20 Mfr # 95-0021-12
COMPARE
60 115 254/365
$568.00USD / Each
Analytik Jena Short/long-wave UV lamp; 4 watts, 254/365 nm wavelength, 230 VAC/50 Hz EW-09817-22 Mfr # 95-0021-10
COMPARE
50 230 254/365
$557.00USD / Each