This product is now discontinued, View Related Products

50-mm In-Line Disc Filters

Simple, quick, and efficient filtration of liquids and gases
Available in a large variety of membrane materials and pore sizes for filtration of liquids and gases. The compact design of the filter capsules reduces hold-up volume and exposure to hazardous chemicals. Use these filter devices in-line or connected to a syringe. Housing is made of polypropylene for excellent chemical compatibility. No adhesives, binders, or surfactants are used in the manufacturing process.
Call For Price

9 variations of this product are available.

See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Membrane Material Pore Size (µm) Sterile
Availability Pricing
ZenPure 50mm in-line disc filter, PTFE, 0.1um, 10/pk EW-49700-70 Mfr # D50CF0102H2H-10
COMPARE
PTFE 0.1
Call For Price
ZenPure 50mm in-line disc filter, PTFE, 0.2um, 10/pk EW-49700-72 Mfr # D50CF0202H2H-10
COMPARE
PTFE 0.2
Call For Price
ZenPure 50mm in-line disc filter, PTFE, 0.45um, 10/pk EW-49700-74 Mfr # D50CF0452H2H-10
COMPARE
PTFE 0.45
Call For Price
ZenPure 50mm in-line disc filter, PTFE, 1.0um, 10/pk EW-49700-76 Mfr # D50CF1002H2H-10
COMPARE
PTFE 1
Call For Price
ZenPure 50mm in-line disc filter, Polyethersufone (PES) with glass microfiber (GMF) 0.2um EW-49700-78 Mfr # D50CS0202H2H-G005-10
COMPARE
Polyethersufone (PES) 0.2
Call For Price
ZenPure In-line disc filter, Polyethersufone (PES) sterile, 0.45um, 10/pk EW-49700-80 Mfr # D50CS0452H2H-G005-ETO-10
COMPARE
Polyethersufone (PES) 0.45
Call For Price
ZenPure In-line disc filter, Nylon w/glass prefilter, 50mm, sterile, 1.2um; 10/Pk EW-49700-82 Mfr # D50CN1202H2H-G005-10
COMPARE
Nylon 1
Call For Price
ZenPure 50mm in-line disc filter, Polypropylene, 5.0um, 10/pk EW-49700-86 Mfr # D50CP0502H2H-10
COMPARE
Polypropylene 5
Call For Price
ZenPure 50mm in-line disc filter, Polypropylene, 10.0um, 10/pk EW-49700-88 Mfr # D50CP1002H2H-10
COMPARE
Polypropylene 10
Call For Price
Showing 1- 9 of 9

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat