This product is now discontinued, View Related Products
500 Series Unmanaged Industrial Ethernet Switches
Ideal for connecting Ethernet-enabled industrial or security equipment
  • Unmanaged switches with full IEEE 802.3 and 1613 compliance
  • RJ-45 ports support full/half duplex operation and auto-sense duplex, speed, and MDIX
  • Redundant power inputs (10 to 30 VDC) with ESD and surge protection diodes on all ports
  • Compact footprint conserves space and is DIN-rail mountable
  • Bi-color LED lights indicate link, speed, activity, and duplex status
  • High MTBF — greater than 2M hours — ideal for harsh conditions
  • Store-and-forward and plug-and-play technology for ease of use
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Input Voltage (VDC) Number of Ports Description
Availability Pricing
Red Lion 509FX N-Tron Unmanaged Industrial Ethernet Switch, 9 Port; SC EW-17702-22 Mfr # 509FX-SC
COMPARE
30 9 N-Tron Unmanaged Industrial Ethernet Switch, 9 Port; SC
Call For Price
Red Lion 509FX-ST N-Tron Unmanaged Industrial Ethernet Switch, 9 Port; ST EW-17702-23 Mfr # 509FX-ST
COMPARE
30 9 N-Tron Unmanaged Industrial Ethernet Switch, 9 Port; ST
Call For Price
Red Lion 516TX N-Tron Unmanaged Industrial Ethernet Switch, 16 Port; TX EW-17702-24 Mfr # 516TX
COMPARE
30 16 N-Tron Unmanaged Industrial Ethernet Switch, 16 Port; TX