A&D Weighing FG-K Series Platform Scales
Fast response time combined with accurate measurement to suit any weighing application
  • One-second response time
  • Stainless steel platform
  • User-selectable filtering function to suit environmental conditions
  • Automatic shutoff extends battery life
  • Parts counting
ITEM(S) ON SALE
$630.80 - $684.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (kg) Capacity (lbs) Power (VAC)
Availability Pricing
A&D Weighing FG-30KAM Platform Scale, 60lb x 0.005 Lb with Display Column EW-11107-01 Mfr # FG-30KAM
COMPARE
30 60 115 / 230
$630.80USD / Each
$760.00USD / Each
A&D Weighing FG-60KAM Platform Scale, 150lb x 0.01 Lb with Display Column EW-11107-11 Mfr # FG-60KAM
COMPARE
60 150 115 / 230
$684.00USD / Each
$760.00USD / Each