Aalborg Stepping Motor Proportioning Valves
Stainless steel models include a 316 SS valve blocks, PTFE-lined aluminum housing blocks, Viton® O-rings, and PFA closing pins. Also includes a 9-pin "D" electrical connector. Optional connection cable (98651-50) for easy input/output wiring. Power input is +12 VDC at 800 mA, protected by a resettable fuse.
$705.00 - $802.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Inlet Port Outlet Port Wetted Materials
Availability Pricing
Aalborg SMV20-A Stepping-Motor Proportioning Valve, Al; 3/8" OD, 1.5-10 VDC EW-98651-00 Mfr # SMV20-A
COMPARE
3/8" compression 3/8" compression Aluminum, Viton, PFA
$705.00USD / Each
Aalborg SMV20-S Stepping-Motor Proportioning Valve, SS; 3/8" OD, 1.5-10 VDC EW-98651-02 Mfr # SMV20-S
COMPARE
3/8" compression 3/8" compression SS, Viton, PFA
$802.00USD / Each
Aalborg SMV30-A Stepping-Motor Proportioning Valve, Al; 1/2" OD, 1.5-10 VDC EW-98651-04 Mfr # SMV30-A
COMPARE
1/2" compression 1/2" compression Aluminum, Viton, PFA
$705.00USD / Each
Aalborg SMV30-S Stepping-Motor Proportioning Valve, SS; 1/2" OD, 1.5-10 VDC EW-98651-06 Mfr # SMV30-S
COMPARE
1/2" compression 1/2" compression SS, Viton, PFA
$802.00USD / Each
Aalborg SMV40-A Stepping-Motor Proportioning Valve, Al; 3/4"NPT(F), 1.5-10 VDC EW-98651-08 Mfr # SMV40-A
COMPARE
3/4" NPT(F) 3/4" NPT(F) Aluminum, Viton, PFA
$705.00USD / Each
Aalborg SMV40-S Stepping-Motor Proportioning Valve, SS; 3/4"NPT(F), 1.5-10 VDC EW-98651-10 Mfr # SMV40-S
COMPARE
3/4" NPT(F) 3/4" NPT(F) SS, Viton, PFA
$802.00USD / Each
Showing 1- 6 of 6
Need Help with this product?
Call Us At1-847-549-7600
Live chat