Abbeon Humidity/Temperature Dial Indicators
Available with a brass or stainless steel case perfect for indoor and outdoor use
  • Large easy-to-read 5" diameter dial
  • Case is drilled for wall mounting
  • No batteries or external power source required
  • Tested at three different humidity positions and includes a certificate of accuracy
These hygrometers are certified to be accurate within ± 3% RH. The dial indicates the complete range of 0 to 100% relative humidity. Each instrument has been tested at three different positions of the dial at temperatures ranging from 32 to 230F. The The large easy-to-read dial. The Durotherm humidity sensor is certified to be accurate within ±3% RH. This sensor is constructed of a man-made fiber that expands and contracts as the humidity changes and mechanically linked through precision bearings and gears to the indicator. A calibration screw is accessible through a slot in the case for periodic calibration. The thermometer features a bimetallic coil sensor and is accurate to 1% of scale.
$293.00 - $330.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
RH Accuracy Max RH Range (%) Description
Availability Pricing
Abbeon HTAB-176 Humidity and Temperature Dial Indicator with White Face and Brass Case EW-37804-58 Mfr # HTAB-176
COMPARE
±3% RH 100 Humidity and Temperature Dial Indicator with White Face and Brass Case
$293.00USD / Each
Abbeon AB-167 Humidity Dial Indicator with Black Face and Brass Case EW-37804-59 Mfr # AB-167
COMPARE
±3% RH 100 Humidity Dial Indicator with Black Face and Brass Case
$304.75USD / Each
Abbeon HTAB-169 Humidity and Temperature Dial Indicator with Black Face and Brass Case
Always in Stock
EW-37804-60 Mfr # HTAB-169
COMPARE
±1% RH 100 Humidity and Temperature Dial Indicator with Black Face and Brass Case
$330.00USD / Each
Abbeon 5063.33 Humidity and Temperature Dial Indicator with White Face and Stainless Steel Case EW-37804-66 Mfr # 5063.33
COMPARE
±3% RH 100 Humidity and Temperature Dial Indicator with White Face and Stainless Steel Case
$307.75USD / Each