Accessories for Cole-Parmer® Vortex Mixers
Improve and expand the performance of you vortex mixer
For use with the variable-speed vortexer, pack contains:
1 x plastic cradle for standard microtitre plate
1 x circular foam insert for beakers and flasks (up to 500ml capacity)
1 x rectangular foam insert (holds 8 x 0.2ml, 8 x 0.5ml and 16 x 1.5ml microcentrifuge tubes)
1 x solid rectangular foam insert (blank for custom holes)
Please note: The microtitre plate holder SA8/1 should not be used at speeds above 800rpm
$96.90USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Vortex Mixer/Shaker Accessory Cup Pack
$96.90USD / Each