This product is now discontinued, View Related Products

Accident Prevention Tags; Danger: Do Not Start

OSHA/ANSI-Compliant Header Tags meet OSHA 29 CFR 1910.145 (f) and ANSI Z535.5 standards which provide specifications, designs, and guidelines for the use and application of accident prevention tags. These tags clearly indicate a temporary hazard that may lead to personal injury or property damage; use until that hazard is eliminated or hazardous operation is complete.

All tags can be written on and have top gabled corners with 3⁄8" reinforced metal grommet on vinyl plastic.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Legend Material
Availability Pricing
Accuform MDT114CTP Tag, Danger Do Not Start, Back A, 5 7/8" X 3 1/8", PF-Cardstock EW-40503-56 Mfr # MDT114CTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! TO DO SO WITHOUT AUTHORITY WILL MEAN DISCIPLINARY ACTION! IT IS HERE FOR A PURPOSE. Cardstock
Call For Price
Accuform MDT114PTP Tag, Danger Do Not Start, Back A, 5 7/8" X 3 3/8", RV-Plastic EW-40503-57 Mfr # MDT114PTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! TO DO SO WITHOUT AUTHORITY WILL MEAN DISCIPLINARY ACTION! IT IS HERE FOR A PURPOSE. Plastic
Call For Price
Accuform MDT190CTP Tag, Danger Do Not Start, Back B, 5 7/8" X 3 1/8", PF-Cardstock EW-40503-58 Mfr # MDT190CTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! Cardstock
Call For Price
Accuform MDT190PTP Tag, Danger Do Not Start, Back B, 5 7/8" X 3 3/8", RV-Plastic EW-40503-59 Mfr # MDT190PTP
COMPARE
Back side: DO NOT REMOVE THIS TAG! Plastic
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat