AccuMac Precision Benchtop RTD Reference Thermometers
High accuracy, fast readings and great stability
AccuMac feature high accuracy, fast readings and great stability.Aailable is dual and single channels models. The dual channelallow measurements and displays of two PRTs simultaneously. All of the readings can be stored in a USB flash drive or transferred to PC through an USB cable. Wireless data transfer can be done with an added optional module.
Both Precision Thermometers allows users to choose ITS-90, IEC-751 (DIN), or Callendar-Van Dusen conversion methods to response to various PRTs. Users can also choose to key in calibration data of each PRT to ensure the best accuracy. They are outstanding choices for precision temperature measurements and calibrations using PRTs, Drywell Calibrators and Temperature Bath.
$1,997.00 - $2,639.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Description
Availability Pricing
AccuMac AM8060 Dual-Channel Precision Benchtop Thermometer EW-90452-71 Mfr # AM8060
COMPARE
1562 Dual-Channel Precision Benchtop Thermometer
$2,639.00USD / Each
AccuMac AM8040 Single-Channel Precision Benchtop Thermometer EW-90452-73 Mfr # AM8040
COMPARE
1562 Single-Channel Precision Benchtop Thermometer
$1,997.00USD / Each