This product is now discontinued, View Related Products
accumet® accuCap™ Electrodes
Most accurate pH electrode, as tested by the PTB (the German Federal Physical Technical Institute in Braunschweig)*
  • Precise, reliable, and fast readings
  • About 200 times larger junction than standard electrodes makes it impossible to clog
The accuCap electrodes use a single pore capillary reference junction, about 200 times larger than typical ceramic junctions. The unique junction design provides increased electrolyte flow, resulting in greater contact between sample and reference, ensuring fast, stable readings. The double junction design offers enhanced performance. A diffusion barrier separates the AgCl reservoir from the reference electrolyte, allowing the use of silver-free electrolytes and preventing AgCl loss due to temperature variations. The end result is an extremely stable reference potential, providing long electrode life.
*See "Traceability of pH measurement" by Petra Spitzer; ISBN 3-89429-877-4 or SSN 0947-7063
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Applications Electrode Body Material Electrode Type
Availability Pricing
Accumet CP620130 accuCap pH Probe, Refillable/DJ/Glass/160x12mm; BNC EW-55500-50 Mfr # CP620130
COMPARE
Refillable Glass Double junction
Call For Price
Accumet CP620131 accuCap pH Probe, Sealed/DJ/Gel-Filled/Glass/130x12mm; BNC EW-55500-51 Mfr # CP620131
COMPARE
Sealed Glass Double junction
Call For Price
Accumet CP620132 accuCap pH Probe, Sealed/DJ/Gel-Filled/Epoxy120x12mm; BNC EW-55500-52 Mfr # CP620132
COMPARE
Sealed Epoxy Double junction
Call For Price
Accumet CP620133 accuCap, glass body, spear tip, gel-filled EW-55500-53 Mfr # CP620133
COMPARE
Spear-tip Glass Double junction