This product is now discontinued, View Related Products

Accumet™ Water Analysis Accessories

Optimize water analysis
  • Choose from a variety of accessories
Replacement cells, electrodes, probes, and meter covers for Accumet™ meters.
ITEM(S) ON SALE
$1,507.90USD / Each

25 variations of this product are available.

See All Product Options Below
25 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Accumet CP-620-155 glass body, non-temperature compensated, 2-cell electrode, K=1 EW-19550-50 Mfr # CP-620-155
COMPARE
glass body, non-temperature compensated, 2-cell electrode, K=1
Call For Price
Accumet CP-620-156 Two-electrode conductivity cell, glass, K=0.1 EW-19550-51 Mfr # CP-620-156
COMPARE
Two-electrode conductivity cell, glass, K=0.1
Call For Price
Accumet CP-620-160 epoxy body, non-temperature compensated, 2-cell electrode, K=1 EW-19550-60 Mfr # CP-620-160
COMPARE
epoxy body, non-temperature compensated, 2-cell electrode, K=1
Call For Price
Accumet CP-620-161 Two-electrode conductivity cell, epoxy, K=0.1 EW-19550-61 Mfr # CP-620-161
COMPARE
Two-electrode conductivity cell, epoxy, K=0.1
Call For Price
Accumet CP-620-162 epoxy body, non-temperature compensated, 2-cell electrode, K=10 EW-19550-62 Mfr # CP-620-162
COMPARE
epoxy body, non-temperature compensated, 2-cell electrode, K=10
Call For Price
Accumet CP-620-101 epoxy body, temperature compensated, 2-cell electrode, K=0.1 EW-19550-63 Mfr # CP-620-101
COMPARE
epoxy body, temperature compensated, 2-cell electrode, K=0.1
Call For Price
Accumet CP-620-100 epoxy body, temperature compensated, 2-cell electrode, K=1 EW-19550-64 Mfr # CP-620-100
COMPARE
epoxy body, temperature compensated, 2-cell electrode, K=1
Call For Price
Accumet CP-620-102 epoxy body, temperature compensated, 2-cell electrode, K=10 EW-19550-65 Mfr # CP-620-102
COMPARE
epoxy body, temperature compensated, 2-cell electrode, K=10
Call For Price
Accumet CP-620-163 Four-electrode conductivity cell, glass, K=1 EW-19550-70 Mfr # CP-620-163
COMPARE
Four-electrode conductivity cell, glass, K=1
Call For Price
Accumet CP-620-164 Four-electrode conductivity cell, glass, K=10.0 EW-19550-71 Mfr # CP-620-164
COMPARE
Four-electrode conductivity cell, glass, K=10.0
Call For Price
Accumet CP-620-165 Four-electrode conductivity cell, epoxy, K=1 EW-19550-72 Mfr # CP-620-165
COMPARE
Four-electrode conductivity cell, epoxy, K=1
Call For Price
Accumet CP-620-166 Four-electrode conductivity cell, epoxy, K=10.0 EW-19550-73 Mfr # CP-620-166
COMPARE
Four-electrode conductivity cell, epoxy, K=10.0
Call For Price
COMPARE
ELECTRODE POLY GEL COMBO
Call For Price
COMPARE
Call For Price
COMPARE
Call For Price
COMPARE
GLASS COMBO RUGGED BULB BNC
Call For Price
COMPARE
PH/TC 3K BNC/SPADELUGS3/4\"CPVC
Call For Price
COMPARE
ELECT DBL JUNC PH/ACT F/AP60S
Call For Price
COMPARE
13-620-300 ELECTRODE
Call For Price
Accumet CP620SSP_01X295703 Replacement Self-Stirring BOD Probe for Meters EW-53109-50 Mfr # CP620SSP_01X295703
COMPARE
Replacement Self-Stirring BOD Probe for Meters
$1,507.90USD / Each
$1,774.00USD / Each
Showing 1 - 20 of 25

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat