Acros Organics Acetone
Quality and consistency your lab can count on
  • Reagents and solutions to meet your lab and research needs
The Acros Organics brand delivers consistent, high-quality organics and fine chemicals at great savings. It also focuses on the evolving trends in organic chemistry, supporting research, development and product activities through a range of off-the-shelf products and expertise in sourcing, manufacturing and technology partnerships. Chemicals sold by Cole-Parmer are not intended for and should not be (i) used for medical, surgical or other patient oriented applications, or (ii) combined with any food product or ingested in any form.
$67.00 - $283.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
Compareitem specifications
Volume (mL) Description
Availability Pricing
Acros Organics Acetone, 99+%, Spectrophotometric Grade; 2.5 L EW-88177-71 Mfr # AC167640025
COMPARE
2500 Acetone, spectrophotometric grade 99+% (2.5 L)
$280.00USD / Each
Acros Organics Acetone, 99+%, Spectrophotometric Grade; 500 mL EW-88177-72 Mfr # AC167645000
COMPARE
500 Acetone, Spectrophotometric grade 99+% (500 mL)
$126.00USD / Each
Acros Organics Acetone, p.a., Liquid; 1 L EW-88189-63 Mfr # AC176800010
COMPARE
1000 Acetone p.a. (1 L) (2-Propanone)
$98.40USD / Each
Acros Organics Acetone, 99+%, Liquid; 2.5 L EW-88190-15 Mfr # AC177170025
COMPARE
2500 Acetone 99+% (2.5 L)
$141.75USD / Each
Acros Organics Acetone, Liquid, HPLC Grade; 1 L EW-88258-90 Mfr # AC268310010
COMPARE
1000 Acetone, for HPLC (1 L)
$118.00USD / Each
Acros Organics Acetone, Water <50 ppm, Extra Dry; 100 mL EW-88294-75 Mfr # AC326801000
COMPARE
100 Acetone, extra dry, water <50 ppm (100 mL)
$157.00USD / Each
Acros Organics Acetone, for Spectroscopy, Reagent ACS; 1 L EW-88337-99 Mfr # AC400100010
COMPARE
1000 Acetone Reagent ACS Spec (1 L)
$115.50USD / Each
Acros Organics Acetone, for Spectroscopy, Reagent ACS; 2.5 L EW-88338-00 Mfr # AC400100025
COMPARE
2500 Acetone Reagent ACS Spec (2.5 L)
$283.00USD / Each
Acros Organics Acetone, 99.5%, for Spectroscopy, Reagent ACS; 500 mL EW-88338-01 Mfr # AC400105000
COMPARE
500 Acetone, reagent ACS, spectro grade 99.5% (500 mL)
$67.00USD / Each
Acros Organics Acetone, 99.5%, Reagent ACS; 500 mL EW-88360-50 Mfr # AC423245000
COMPARE
500 Acetone, reagent ACS 99.5% (500 mL)
$116.50USD / Each
Showing 1- 10 of 10